Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563801 Supplier Invitrogen™ Supplier No. PA563801
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEPQHEMVAH RHRQEVLCKE MVPLAEQTPL TLQSQPKEPQ LTCDSAQKCH SIGETDEVTK T Highest antigen sequence identity to the following orthologs: Mouse - 31%, Rat - 29%.

ZNF193 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 5 C2H2-type zinc fingers and 1 SCAN box domain. ZNF193 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZSCAN9
Gene Accession No. O15535
Gene Alias Cell proliferation-inducing gene 12 protein; cell proliferation-inducing protein 12; PIG12; PRD51; zinc finger and SCAN domain containing 9; zinc finger and SCAN domain-containing protein 9; Zinc finger protein 193; ZNF193; ZSCAN9
Gene Symbols ZSCAN9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZSCAN9. Recombinant protein control fragment (Product #RP-101311).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7746
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.