Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSCAN9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112793
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112793 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112793 Supplier Invitrogen™ Supplier No. PA5112793
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZSCAN9 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ZNF193 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 5 C2H2-type zinc fingers and 1 SCAN box domain. ZNF193 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZSCAN9
Gene Accession No. O15535
Gene Alias Cell proliferation-inducing gene 12 protein; cell proliferation-inducing protein 12; PIG12; PRD51; zinc finger and SCAN domain containing 9; zinc finger and SCAN domain-containing protein 9; Zinc finger protein 193; ZNF193; ZSCAN9
Gene Symbols ZSCAN9
Host Species Rabbit
Immunogen Recombinant Human Zinc finger and SCAN domain-containing protein 9 protein (1-394AA).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7746
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.