Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZsGreen1 Mouse anti-Tag, Clone: OTI2C2 (formerly 2C2), TrueMAB™, Origene

Mouse Monoclonal Antibody
Supplier: OriGene CF180002

This item is not returnable.
View return policy
Description
ZsGreen1 Monoclonal antibody specifically detects ZsGreen1 in Tag samples. It is validated for Western BlotSpecifications
ZsGreen1 | |
Monoclonal | |
Unconjugated | |
ZsGreen1 | |
This antibody is raised against a recombinant protein of ZsGreen1 expressed in HEK293 cells. | |
100 μg | |
Primary | |
Antibody | |
IgG1 |
Western Blot | |
OTI2C2 (formerly 2C2) | |
PBS with 8% trehalose and no preservative | |
Mouse | |
Protein A/G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized | |
Tag |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction