Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZSWIM2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531253
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531253 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531253 Supplier Invitrogen™ Supplier No. PA531253
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Human (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

E3 ubiquitin-protein ligase involved in the regulation of Fas-, DR3- and DR4-mediated apoptosis. Functions in conjunction with the UBE2D1, UBE2D3 and UBE2E1 E2 ubiquitin-conjugating enzymes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSWIM2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZSWIM2
Gene Accession No. Q8NEG5
Gene Alias E3 ubiquitin-protein ligase ZSWIM2; MEKK1-related protein X; MEX; RING-type E3 ubiquitin transferase ZSWIM2; Zinc finger SWIM domain-containing protein 2; zinc finger SWIM-type containing 2; zinc finger, SWIM domain containing 2; zinc finger, SWIM-type containing 2; ZSWIM2; ZZ-type zinc finger-containing protein 2; ZZZ2
Gene Symbols ZSWIM2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZSWIM2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.