Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZSWIM3 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8846 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8846 Supplier Bioss Supplier No. BS9138R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ZSWIM3 is a protein coding gene. Gene Ontology (GO) annotations related to this gene include zinc ion binding.

Specifications

Antigen ZSWIM3
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZSWIM3
Gene Accession No. Q96MP5
Gene Alias 4921517A06Rik; C20orf164; C86566; LOC100339103; PPP1R174; protein phosphatase 1, regulatory subunit 174; Zinc finger SWIM domain-containing protein 3; zinc finger SWIM-type containing 3; zinc finger, SWIM domain containing 1-like; zinc finger, SWIM domain containing 3; zinc finger, SWIM-type containing 3; ZSWIM3
Gene Symbols ZSWIM3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZSWIM3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140831
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.