Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZUFSP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560709
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560709 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560709 Supplier Invitrogen™ Supplier No. PA560709
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RINTVQYGTS DNKKDNTLQC GMEVNSSILS GCASNHPKNS AQNLTKDSTL KHEGFYSENL TESRKFLKSR EKQSSLTEIK GSVYETTYSP PECPFCGKIE EHSEDMETHV KTKHANLLDI PLEDCDQPLY DCPMC Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 64%.

Deubiquitinase with endodeubiquitinase activity that specifically interacts with and cleaves 'Lys-63'-linked long polyubiquitin chains. Shows only weak activity against 'Lys-11' and 'Lys-48'-linked chains. Plays an important role in genome stability pathways, functioning to prevent spontaneous DNA damage and also promote cellular survival in response to exogenous DNA damage. Modulates the ubiquitination status of replication protein A (RPA) complex proteins in response to replication stress.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZUFSP
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZUFSP
Gene Accession No. Q96AP4
Gene Alias 2700019D07Rik; C6orf113; dJ412I7.3; DUB; Lys-63-specific deubiquitinase ZUFSP; zinc finger with UFM1 specific peptidase domain; zinc finger with UFM1-specific peptidase domain; zinc finger with UFM1-specific peptidase domain protein; Zinc finger-containing ubiquitin peptidase 1; Zufsp; Zup1
Gene Symbols ZUFSP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZUFSP. Recombinant protein control fragment (Product #RP-88721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221302
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.