Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZUFSP Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5145149
Description
Immunogen sequence: TSKPPIYLQH QGHSRTVIGI EEKKNRTLCL LILDPGCPSR EMQKLLKQDI EASSLKQLRK SMGNLKHKQY QILAVEGALS LEEKLARRQA SQVFTA Antigen sequence identity: Mouse 89%; Rat 84%.
Deubiquitinase with endodeubiquitinase activity that specifically interacts with and cleaves 'Lys-63'-linked long polyubiquitin chains. Shows only weak activity against 'Lys-11' and 'Lys-48'-linked chains. Plays an important role in genome stability pathways, functioning to prevent spontaneous DNA damage and also promote cellular survival in response to exogenous DNA damage. Modulates the ubiquitination status of replication protein A (RPA) complex proteins in response to replication stress.
Specifications
ZUFSP | |
Polyclonal | |
Unconjugated | |
ZUFSP | |
2700019D07Rik; C6orf113; dJ412I7.3; DUB; Lys-63-specific deubiquitinase ZUFSP; zinc finger with UFM1 specific peptidase domain; zinc finger with UFM1-specific peptidase domain; zinc finger with UFM1-specific peptidase domain protein; Zinc finger-containing ubiquitin peptidase 1; Zufsp; Zup1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
221302 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96AP4 | |
ZUFSP | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 479-574. Recombinant protein control fragment (Product #RP-109915). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction