Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZW10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562357
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562357 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562357 Supplier Invitrogen™ Supplier No. PA562357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSTVPLAEML GDMIWEDLSE CLIKNCLVYS IPTNSSKLQQ YEEIIQSTEE FENALKEMRF LKGDTTDLLK YARNINSHFA NKKCQDVIVA ARNLMTSE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.

This gene encodes a protein that is one of many involved in mechanisms to ensure proper chromosome segregation during cell division.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZW10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZW10
Gene Accession No. O43264
Gene Alias 6330566F14Rik; centromere/kinetochore protein zw10; centromere/kinetochore protein zw10 homolog; H KNTC1AP; HZW10; KNTC1AP; LOW QUALITY PROTEIN: centromere/kinetochore protein zw10 homolog; MGC149821; MmZw10; Unknown (protein for MGC:139579); zeste white 10 homolog; Zw10; ZW10 homolog, centromere/kinetochore protein; zw10 kinetochore protein; ZW10, kinetochore associated, homolog
Gene Symbols ZW10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZW10. Recombinant protein control fragment (Product #RP-103093).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9183
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.