Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZWILCH Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989298
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9298 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9298 Supplier Bioss Supplier No. BS9732R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Essential component of the mitotic checkpoint, which prevents cells from prematurely exiting mitosis. Required for the assembly of the dynein-dynactin and MAD1-MAD2 complexes onto kinetochores.

Specifications

Antigen ZWILCH
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ZWILCH
Gene Accession No. Q9H900
Gene Alias 2310031L18Rik; homolog of Drosophila Zwilch; hZwilch; KNTC1AP; Protein zwilch homolog; Zwilch; zwilch kinetochore protein; Zwilch, kinetochore associated, homolog
Gene Symbols ZWILCH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human ZWILCH.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55055
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.