Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZWINT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA592903
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA592903 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA592903 Supplier Invitrogen™ Supplier No. PA592903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEAAETEAEA AALEVLAEVA GILEPVGLQE EAELPAKILV EFVVDSQKKD KLLCSQLQVA DFLQNILAQE DTAKGLDPLA SEDTSRQKAI AAKEQWKELK ATYREHVEAI KIGLTKALTQ MEEAQRKRTQ LREAFEQLQA KKQMAMEKRR AVQNQWQLQQ EKHLQHLAEV SAEVRERKTG TQQELDRVFQ KLGNLKQQAE QERDKLQRYQ TFLQLLYTLQ GKLLFPEAEA EAENLPDDKP QQPTRPQEQS TGDTMGRDPG VSFKAVGLQP AGDVNLP; Positive Samples: 293T, SW620, HepG2, Jurkat, Mouse brain; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.

This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZWINT
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.01 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZWINT
Gene Accession No. O95229, Q8VIL3, Q9CQU5
Gene Alias 2010007E07Rik; 2600001N01Rik; D10Ertd749e; human ZW10 interacting protein-1; HZwint-1; KNTC2AP; neuroprotective protein 8; scoilin; Sip30; SNAP25 interacting protein 30; SNAP25 interacting protein of 30 kDa; SNAP25-interacting protein 30; ZW10 interacting kinetochore protein; ZW10 interacting protein-1; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; ZWINT; ZWINT1; Zwint-1
Gene Symbols ZWINT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human ZWINT (NP_0089882).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11130, 257644, 52696
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.