Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZXDA Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP204129FR
Description
ZXDA Polyclonal antibody specifically detects ZXDA in Human samples. It is validated for Western BlotSpecifications
ZXDA | |
Polyclonal | |
Western Blot | |
zinc finger X-linked protein ZXDA, zinc finger, X-linked, duplicated A, ZNF896 | |
The immunogen recognized by this antibody maps to this antibody and Zinc Finger, X-linked, Duplicated B using the numbering given in entry NP_009087.1 and NP_009088.1 (GeneID 7789 and 158586). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
7789 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction