Learn More
Invitrogen™ ZXDA Polyclonal Antibody

Description
ZXDA Polyclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | ZXDA |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.8 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ZXDA |
| Gene Accession No. | P98168 |
| Gene Alias | zinc finger protein 896; zinc finger X-linked protein ZXDA; zinc finger, X-linked, duplicated A; ZNF896; ZXDA |
| Gene Symbols | ZXDA |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.