Learn More
Invitrogen™ ZXDB Polyclonal Antibody

Description
Immunogen sequence: DPPQSLDTSL FFGTAATGFQ QSSLNMDEVS SVSVGPLGSL DSLAMKNSSP EPQAL Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 42%.
Specifications
Specifications
| Antigen | ZXDB |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZXDB |
| Gene Accession No. | P98169 |
| Gene Alias | dJ83L6.1; Zinc finger X-linked protein ZXDB; zinc finger, X-linked, duplicated B; ZNF896; ZNF905; ZXDB |
| Gene Symbols | ZXDB |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.