Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZXDC Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565568
Description
Immunogen sequence: KIKEGKMSPPH FHASQNSWLC GSLVVPSGGR PGPAPAAGVQ CGAQGVQVQL VQDDPSGEGV LPSARGPATF LPFLTVDLP Highest antigen sequence identity to the following orthologs - mouse 55%, rat 52%.
ZXDC belongs to the ZXD family. It contains 10 C2H2-type zinc fingers. ZXDC cooperates with CIITA to promote transcription of MHC class I and MHC class II genes.
Specifications
ZXDC | |
Polyclonal | |
Unconjugated | |
ZXDC | |
A930012H20; B930086F11Rik; BC003332; LOW QUALITY PROTEIN: zinc finger protein ZXDC; RGD1561296; SERH2790; zinc finger protein ZXDC; ZXD family zinc finger C; ZXD family zinc finger C-like; ZXDC; ZXDL; ZXD-like zinc finger protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79364 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2QGD7 | |
ZXDC | |
Recombinant Human ZXDC. Recombinant protein control fragment (Product #RP-104875). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction