Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZYG11A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565065 Supplier Invitrogen™ Supplier No. PA565065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSREMEVSYFA AGIIAHLTSD RQLWISRDFQ RRTLLQDLHA TIQNWPSSSC KMTALVTYRS FKTFFPLLGN FSQPEVQLWA L Highest antigen sequence identity to the following orthologs - mouse 68%, rat 72%.

Probably acts as target recruitment subunit in an E3 ubiquitin ligase complex ZYGA-CUL2-elongin BC.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZYG11A
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZYG11A
Gene Accession No. Q6WRX3
Gene Alias protein zyg-11 homolog A; ZYG11; zyg-11 family member A, cell cycle regulator; zyg-11 homolog A; ZYG11A; ZYG-11A early embryogenesis protein
Gene Symbols ZYG11A
Host Species Rabbit
Immunogen Recombinant Human ZYG11A. Recombinant protein control fragment (Product #RP-105828).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 440590
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.