Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZYX Mouse anti-Human, Mouse, Unconjugated, Clone: 2D1, Abnova™

Catalog No. 89335030 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-030 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-030 Supplier Abnova Corporation Supplier No. H00007791M02J
Add to Cart
Add to Cart

Mouse Monoclonal Antibody.

ZYX Monoclonal antibody specifically detects ZYX in Human, Mouse samples. It is validated for IHC-P, S-ELISA, ELISA, WB-Re.

Specifications

Antigen ZYX
Applications Immunohistochemistry (Paraffin), ELISA, Western Blot
Classification Monoclonal
Clone 2D1
Conjugate Unconjugated
Description zyxin
Gene ZYX
Gene Accession No. BC008743.2
Gene Alias ESP-2, HED-2
Host Species Mouse
Immunogen ZYX (AAH08743.1, 1 a.a. ∼ 572 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 7791
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.