Learn More
Invitrogen™ Zyxin Polyclonal Antibody

Description
Positive test controls include: HeLa. The target is usually found in the following locations: Cell junction, Cytoplasm, Nucleus, cytoskeleton, focal adhesion. Immunogen sequence: PLTLKEVEEL EQLTQQLMQD MEHPQRQNVA VNELCGRCHQ PLARAQPAVR ALGQLFHIAC FTCHQCAQQL QGQQFYSLEG APYCEGCYTD TLEKCNTCGE PITDRMLRAT GKAYHPHCFT CVVCARPLEG TSFIVDQANR PHCVPDYHKQ YAPRCSVCSE PIMPEPGRDE TVRVVALDKN FHMKCYKCED CGKPLSIEAD DNGCFPLDGH VLCRKCHTAR AQT.
Specifications
Specifications
| Antigen | Zyxin |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.25 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | ZYX |
| Gene Accession No. | Q15942, Q62523 |
| Gene Alias | 9530098H06Rik; ESP-2; HED-2; LOW QUALITY PROTEIN: zyxin; R75157; ZYX; Zyxin; zyxin-2 |
| Gene Symbols | ZYX |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.