Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zyxin Rabbit anti-Human, Mouse, Clone: 4F9P3, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Zyxin |
---|---|
Clone | 4F9P3 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Zyxin Monoclonal antibody specifically detects Zyxin in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Zyxin | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
7791 | |
IgG | |
Affinity purified |
4F9P3 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
CTCL tumor antigen se14-3, CTCL-associated antigen se14-3, cutaneous T-cell lymphoma associated antigen se14-3, Cutaneous T-cell lymphoma-associated antigen se14-3, KIAA1125, MGC31836, predicted protein of HQ2893, PRKCBP1, PRO2893, protein kinase C-binding protein 1, Rack7, RACK7protein kinase C binding protein 1, zinc finger MYND domain containing protein 8, Zinc finger MYND domain-containing protein 8, zinc finger, MYND-type containing 8 | |
A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Zyxin (Q15942). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title