Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Zyxin Recombinant Rabbit Monoclonal Antibody (4F9P3)
GREENER_CHOICE

Catalog No. PIMA542774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542774 Supplier Invitrogen™ Supplier No. MA542774
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive test controls include: MCF7, Mouse lung. The target is usually found in the following locations: Cell junction, Cytoplasm, Nucleus, cytoskeleton, focal adhesion. Immunogen sequence: TQPRGPPASS PAPAPKFSPV TPKFTPVASK FSPGAPGGSG SQPNQKLGHP EALSAGTGSP QPPSFTYAQQ REKPRVQEKQ HPVPPPAQNQ NQVRSPGAPG P.

Zyxin is involved in focal adhesions, which are actin-rich structures that enable cells to adhere to the extracellular matrix and at which protein complexes involved in signal transduction assemble. Zyxin is a zinc-binding phosphoprotein that concentrates at focal adhesions and along the actin cytoskeleton. Zyxin has an N-terminal proline-rich domain and three LIM domains in its C-terminal half. The proline-rich domain may interact with SH3 domains of proteins involved in signal transduction pathways while the LIM domains are likely involved in protein-protein binding. Zyxin may function as a messenger in the signal transduction pathway that mediates adhesion-stimulated changes in gene expression and may modulate the cytoskeletal organization of actin bundles. Alternative splicing results in multiple transcript variants that encode the same isoform.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Zyxin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4F9P3
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ZYX
Gene Accession No. Q15942, Q62523
Gene Alias 9530098H06Rik; ESP-2; HED-2; LOW QUALITY PROTEIN: zyxin; R75157; ZYX; Zyxin; zyxin-2
Gene Symbols ZYX
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human Zyxin (Q15942).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22793, 7791
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.