Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZZEF1 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232119APC
Description
ZZEF1 Polyclonal antibody specifically detects ZZEF1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)Specifications
ZZEF1 | |
Polyclonal | |
Immunoprecipitation, Western Blot (Negative) | |
FLJ10821, FLJ23789, KIAA0399, MGC166929, zinc finger, ZZ-type with EF hand domain 1, zinc finger, ZZ-type with EF-hand domain 1, ZZZ4 | |
The immunogen this antibody was made to, maps to a region between residue 525 to 575 of human Zinc Finger, ZZ-Type with EF-Hand Domain 1 using the numbering given in entry NP_055928.3 (GeneID 23140). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunoprecipitation | |
APC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23140 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction