Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZZEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556856
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556856 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556856 Supplier Invitrogen™ Supplier No. PA556856
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DYFFLEVQKR FDGDELTTDE RIRSLAQRWQ PSKSLRLEEQ SAKAVDTDMI ILPCLSRPAR CDQATAESNP VTQKLI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

ZZEF1 gene ontology annotations related to this gene include calcium ion binding; zinc ion binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZZEF1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZZEF1
Gene Accession No. O43149
Gene Alias FLJ10821; KIAA0399; zinc finger ZZ-type and EF-hand domain containing 1; zinc finger ZZ-type and EF-hand domain-containing protein 1; zinc finger, ZZ-type with EF hand domain 1; zinc finger, ZZ-type with EF-hand domain 1; ZZEF1; ZZZ4
Gene Symbols ZZEF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZZEF1. Recombinant protein control fragment (Product #RP-95571).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23140
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.