Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZZEF1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566814
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6814 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6814 Supplier Bethyl Laboratories, Inc Supplier No. A304069AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

ZZEF1 gene ontology annotations related to this gene include calcium ion binding; zinc ion binding.

Specifications

Antigen ZZEF1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ZZEF1
Gene Accession No. O43149
Gene Alias FLJ10821, KIAA0399, zinc finger, ZZ-type with EF hand domain 1, zinc finger, ZZ-type with EF-hand domain 1, ZZZ4
Gene Symbols ZZEF1
Host Species Rabbit
Immunogen Between 1900 and 1950
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23140
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.