Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZZZ3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101968 Supplier Invitrogen™ Supplier No. PA5101968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ZZZ3.

Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZZZ3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ZZZ3
Gene Accession No. Q6KAQ7, Q8IYH5
Gene Alias 3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC component 1 homolog; ATAC1; AW556059; DKFZP564I052; R75514; RGD1565468; zinc finger ZZ-type containing 3; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3; Zzz3
Gene Symbols ZZZ3
Host Species Rabbit
Immunogen A synthesized peptide derived from human ZZZ3(Accession Q8IYH5), corresponding to amino acid residues Y199-T249.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108946, 26009, 310958
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.