Filtered Search Results
Fisherbrand™ Powder Free Nitrile Gloves
Choose a latex-free glove with exceptional durability and puncture resistance. Fisherbrand™ powder-free Nitrile Exam Gloves provide a superior fit, comfort, tactile sensitivity and dexterity.
| Disposable | Single-use |
|---|---|
| Length (Metric) | 245 mm |
| Applications | Healthcare,Industrial,Laboratory |
| Chemotherapeutic Rated | No |
| Finger Thickness (English) | 4.7 mil |
| Color | Blue |
| Length (English) | 9.6 in. |
| Quantity | 100 Gloves/Box |
| Material | Nitrile |
| Powdered | Powder Free |
| Sterility | Non-sterile |
| Finger Thickness (Metric) | 0.12 mm |
| Thickness (Metric) | Palm: 0.09 mm, Finger: 0.12 mm |
| FDA 510 (k) | Yes |
| Thickness (English) | Palm: 3.5 mil, Finger: 4.7 mil |
| Palm Thickness (English) | 3.5 mil |
| Latex Free | Yes |
| Exterior Finish | Fully Textured |
| Certifications/Compliance | ASTM D6319,FDA21 CFR 177-2600-US Food Contact Approved,US QSR/FDA 510(k) Medical Examination Grade |
| AQL | 1.5 |
| Palm Thickness (Metric) | 0.09 mm |
Fisherbrand™ Reusable Glass Low-Form Griffin Beakers
Small and Specialty Supplier Partner
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Small and/or specialty supplier based on Federal laws and SBA requirements.
Learn More
Made of KG-33 borosilicate glass
| Disposable | Reusable |
|---|---|
| Includes | Spout |
| Type | Beaker |
| Description | Beaker |
| Style | Griffin |
| Graduated | Yes |
| Scale | Dual Scale |
| Material | Borosilicate Glass |
| Certifications/Compliance | ASTM E960, ASTM E438, USP Type I |
| Label Type | Matte Finish Marking |
| For Use With (Application) | Excellent Mechanical Strength and Durability, While Providing High Resistance to Chemical Attack and Thermal Shock |
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
CAS: 64-17-5 Molecular Formula: C2H6O InChI Key: LFQSCWFLJHTTHZ-UHFFFAOYSA-N Synonym: ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol PubChem CID: 702 ChEBI: CHEBI:16236 IUPAC Name: ethanol SMILES: CCO
| PubChem CID | 702 |
|---|---|
| CAS | 64-17-5 |
| ChEBI | CHEBI:16236 |
| SMILES | CCO |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| IUPAC Name | ethanol |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| Molecular Formula | C2H6O |
Fisherbrand™ Transfer Pipettes
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Disposable transfer pipette for general purpose use. Manufactured with IVD designation (FDA Class I, Product Code: JRC).
| Disposable | Single-use |
|---|---|
| Color | Natural |
| Sterility | Non-sterile |
| Height (Metric) | 15 mm |
|---|---|
| Type | Petri Dish |
| Shape | Round |
| Material | Polystyrene |
| Closure Color | Clear |
| Sterility | Sterile |
| Closure Included | Closure Included |
Water, (DNASE, RNASE free), Fisher BioReagents™
CAS: 7732-18-5 Molecular Formula: H2O Molecular Weight (g/mol): 18.015 InChI Key: XLYOFNOQVPJJNP-UHFFFAOYSA-N Synonym: dihydrogen oxide,dihydrogen monoxide PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: oxidane SMILES: O
| PubChem CID | 962 |
|---|---|
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18.015 |
| ChEBI | CHEBI:15377 |
| SMILES | O |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| IUPAC Name | oxidane |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Molecular Formula | H2O |
Water, Optima™ LC/MS Grade, Fisher Chemical™
CAS: 7732-18-5 Molecular Formula: H2O Molecular Weight (g/mol): 18.015 MDL Number: 11332 InChI Key: XLYOFNOQVPJJNP-UHFFFAOYSA-N Synonym: dihydrogen oxide,dihydrogen monoxide PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: oxidane SMILES: O
| PubChem CID | 962 |
|---|---|
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18.015 |
| ChEBI | CHEBI:15377 |
| MDL Number | 11332 |
| SMILES | O |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| IUPAC Name | oxidane |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Molecular Formula | H2O |
Water, Molecular Biology Grade, Fisher BioReagents
CAS: 7732-18-5 Molecular Formula: H2O Molecular Weight (g/mol): 18.015 InChI Key: XLYOFNOQVPJJNP-UHFFFAOYSA-N Synonym: dihydrogen oxide,dihydrogen monoxide PubChem CID: 962 ChEBI: CHEBI:15377 IUPAC Name: oxidane SMILES: O
| PubChem CID | 962 |
|---|---|
| CAS | 7732-18-5 |
| Molecular Weight (g/mol) | 18.015 |
| ChEBI | CHEBI:15377 |
| SMILES | O |
| Synonym | dihydrogen oxide,dihydrogen monoxide |
| IUPAC Name | oxidane |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Molecular Formula | H2O |
Acetone (Certified ACS), Fisher Chemical™
CAS: 67-64-1 Molecular Formula: C3H6O Molecular Weight (g/mol): 58.08 MDL Number: MFCD00008765 InChI Key: CSCPPACGZOOCGX-UHFFFAOYSA-N Synonym: acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone PubChem CID: 180 ChEBI: CHEBI:15347 IUPAC Name: propan-2-one SMILES: CC(C)=O
| PubChem CID | 180 |
|---|---|
| CAS | 67-64-1 |
| Molecular Weight (g/mol) | 58.08 |
| ChEBI | CHEBI:15347 |
| MDL Number | MFCD00008765 |
| SMILES | CC(C)=O |
| Synonym | acetone,2-propanone,propanone,dimethyl ketone,methyl ketone,dimethylformaldehyde,pyroacetic ether,beta-ketopropane,dimethylketal,chevron acetone |
| IUPAC Name | propan-2-one |
| InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
| Molecular Formula | C3H6O |
Dimethyl Sulfoxide, Fisher BioReagents™
CAS: 67-68-5 Molecular Formula: C2H6OS Molecular Weight (g/mol): 78.13 MDL Number: MFCD00002089 InChI Key: IAZDPXIOMUYVGZ-UHFFFAOYSA-N Synonym: dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide PubChem CID: 679 ChEBI: CHEBI:28262 IUPAC Name: methylsulfinylmethane SMILES: CS(C)=O
| PubChem CID | 679 |
|---|---|
| CAS | 67-68-5 |
| Molecular Weight (g/mol) | 78.13 |
| ChEBI | CHEBI:28262 |
| MDL Number | MFCD00002089 |
| SMILES | CS(C)=O |
| Synonym | dimethyl sulfoxide,dmso,methyl sulfoxide,dimethylsulfoxide,dimethyl sulphoxide,methane, sulfinylbis,demsodrox,demasorb,demavet,dimexide |
| IUPAC Name | methylsulfinylmethane |
| InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
| Molecular Formula | C2H6OS |
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
CAS: 64-18-6 Molecular Formula: CH2O2 InChI Key: BDAGIHXWWSANSR-UHFFFAOYSA-N Synonym: methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt PubChem CID: 284 ChEBI: CHEBI:30751 IUPAC Name: formic acid
| PubChem CID | 284 |
|---|---|
| CAS | 64-18-6 |
| ChEBI | CHEBI:30751 |
| Synonym | methanoic acid,formylic acid,aminic acid,bilorin,hydrogen carboxylic acid,formisoton,myrmicyl,formira,acide formique,collo-bueglatt |
| IUPAC Name | formic acid |
| InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
| Molecular Formula | CH2O2 |
Fisherbrand™ Maxima Rotary Vane Vacuum Pumps - 115 V, 60 Hz
Fisherbrand™ Maxima rotary vane vacuum pumps extend service intervals due to robust materials and design features.
| Includes | Exhaust Filter, Oil Fill Funnel, Hose Clamp |
|---|---|
| Enclosure Rating | IP54 |
| Electrical Requirements | 115 V, 60 Hz, 1 Ph, N.A. 115 V Plug |
| Applications | Vacuum Manifolds, Vacuum Oven, Mass Spectroscopy, Backing Turbo Pumps, Sputter Coater, Tube Furnace |
| Voltage | 115 V |
Ammonium Hydroxide (Certified ACS Plus), Fisher Chemical™
CAS: 1336-21-6 Molecular Formula: H5NO Molecular Weight (g/mol): 35.05 MDL Number: MFCD00066650 InChI Key: VHUUQVKOLVNVRT-UHFFFAOYSA-N Synonym: ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh PubChem CID: 14923 ChEBI: CHEBI:18219 IUPAC Name: amine hydrate SMILES: N.O
| PubChem CID | 14923 |
|---|---|
| CAS | 1336-21-6 |
| Molecular Weight (g/mol) | 35.05 |
| ChEBI | CHEBI:18219 |
| MDL Number | MFCD00066650 |
| SMILES | N.O |
| Synonym | ammonium hydroxide,ammonia water,ammonia aqueous,aquammonia,aqua ammonia,ammonia, aqua,household ammonia,ammonium hydroxide solution,ammonia, monohydrate,ammonium hydroxide nh4 oh |
| IUPAC Name | amine hydrate |
| InChI Key | VHUUQVKOLVNVRT-UHFFFAOYSA-N |
| Molecular Formula | H5NO |
Hexanes (Certified ACS), Fisher Chemical™
CAS: 92112-69-1 Molecular Formula: C6H14 Molecular Weight (g/mol): 86.18 MDL Number: MFCD02179311 InChI Key: VLKZOEOYAKHREP-UHFFFAOYSA-N Synonym: n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes PubChem CID: 8058 ChEBI: CHEBI:29021 IUPAC Name: hexane SMILES: CCCCCC
| PubChem CID | 8058 |
|---|---|
| CAS | 92112-69-1 |
| Molecular Weight (g/mol) | 86.18 |
| ChEBI | CHEBI:29021 |
| MDL Number | MFCD02179311 |
| SMILES | CCCCCC |
| Synonym | n-hexane,esani,skellysolve b,gettysolve-b,hexyl hydride,hexan,dipropyl,heksan,hexanen,hexanes |
| IUPAC Name | hexane |
| InChI Key | VLKZOEOYAKHREP-UHFFFAOYSA-N |
| Molecular Formula | C6H14 |
Fisherbrand™ Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Greener Choice
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Offers one or more environmental benefits itemized in the U.S. FTC Green Guides.
Learn More
Pipets have improved optical clarity — high-contrast black markings make volumes easier to read
| Disposable | Single-use |
|---|---|
| Graduated | Yes |
| Material | Polystyrene |
| Certifications/Compliance | Non-pyrogenic, USP XXII, USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free |