H-B Instrument Education Products
Certified, lab-grade, pocket and select special application type thermometers
Features:
- Green EnviroKleen-certified, environmentally friendly Enviro-Safe liquid and recyclable SafetyPak® packaging
- White, lead-free glass
- -20° to +260°C
- 0° to 300°F
Certified, lab-grade thermometers
Features:
- Black EnviroKleen-certified, environmentally friendly Enviro-Safe liquid and recyclable SafetyPak packaging
- High-contrasting black liquid against yellow back glass makes this thermometer the easiest to read liquid-in-glass thermometer on the market
- -20° to +260°C
- 0° to 300°F
Lab-grade thermometers
Features:
- PFA Coated Lab-grade thermometers
- Black EnviroKleen-certified, environmentally friendly Enviro-Safe liquid and recyclable SafetyPak packaging
- High-contrasting black liquid against yellow back glass makes this thermometer the easiest to read liquid-in-glass thermometer on the market
- -20° to +150°C
- 0° to 300°F
Verification thermometers (found in bottles featuring liquid and in the thermometers of both Easy-Read and Enviro-Safe styles)
Features:
- PFA-safety coating
- Recyclable packaging
- -90° to +200°C
- 20° to 60°F
HB Instruments Calibration
HB Instruments Instructional Video reviews the terminology and documentation supplied with the instruments.
HB Instruments Reuinite
HB Instruments Instructional Video includes two methods to reunite your glass themometer.
HB Instruments Thermometer
HB Instruments Instructional Video reviws various types of liquids used in liquid themometers.
HB Instruments Immersion
HB Instruments Instructional Video reviews the use of partial, total or complete immersion themometers.
About H-B Instrument
About H-B Instrument, an SP Scienceware brand, is known worldwide as a quality manufacturer of temperature and density measuring instruments. Its Enviro-Safe thermometer liquid is 100% non-toxic, non-hazardous and biodegradable. Better yet, it cleans up with soap and water! Enviro-Safe liquid is the only EnviroKleen™-certified environmentally friendly thermometer liquid available on the market.
All instruments are tested in H-B's exclusive triple accredited/registered ISO/IEC 9001:2008, ISO/IEC 14001:2004, ISO/IEC 17025:2005 facility and A2LA Accredited Calibration Laboratory (Certificate #2448.01); internationally recognized and accepted by the signatory economies of the ILAC/MRA. They are packaged with a multi-language (English, French, German, Italian, Portuguese and Spanish) Statement of Accuracy indicating accuracy traceable to NIST.