CAS RN 103321-50-2
CAS RN 103321-50-2
IUPAC Name:
(9H-fluoren-9-yl)methyl N-[(2S)-1-chloro-1-oxopropan-2-yl]carbamate
Synonyms:
9H-fluoren-9-ylmethyl N-[(2S)-1-chloro-1-oxopropan-2-yl]carbamate
Molecular Weight (g/mol):
329.78
Molecular Formula:
C18H16ClNO3
InChi Key:
AFMYCMGTXVCJCH-NSHDSACASA-N
SMILES:
C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(Cl)=O
1
–
1
of
1
results
1
Advanced Chem Tech Fmoc-Ala-Cl, Advanced ChemTech
CAS: 103321-50-2 Molecular Formula: C18H16ClNO3 Molecular Weight (g/mol): 329.78 MDL Number: MFCD00235802 InChI Key: AFMYCMGTXVCJCH-NSHDSACASA-N IUPAC Name: (9H-fluoren-9-yl)methyl N-[(2S)-1-chloro-1-oxopropan-2-yl]carbamate SMILES: C[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(Cl)=O