CAS RN 104104-50-9
IUPAC Name:
hydrogen 4-[6-imino-3-(4-methoxyphenyl)-1,6-dihydropyridazin-1-yl]butanoic acid bromide
Synonyms:
gabexate
Molecular Weight (g/mol):
368.23
Molecular Formula:
C15H18BrN3O3
InChi Key:
GFZHNFOGCMEYTA-UHFFFAOYSA-N
SMILES:
[H+].[Br-].COC1=CC=C(C=C1)C1=NN(CCCC(O)=O)C(=N)C=C1
1
–
1
of
1
results
1
R&D Systems™ SR 95531 hydrobromide
CAS: 104104-50-9 Molecular Formula: C15H18BrN3O3