CAS RN 1051487-82-1
CAS RN 1051487-82-1
IUPAC Name:
N'-[1-(3-bromophenyl)ethylidene]-2-oxo-4-phenylpyrrolidine-3-carbohydrazide
Synonyms:
N'-[1-(3-bromophenyl)ethylidene]-2-oxo-4-phenylpyrrolidine-3-carbohydrazide
Molecular Weight (g/mol):
400.28
Molecular Formula:
C19H18BrN3O2
InChi Key:
RQKXQCSEZPQBNZ-UHFFFAOYNA-N
SMILES:
CC(=NNC(=O)C1C(CNC1=O)C1=CC=CC=C1)C1=CC(Br)=CC=C1
1
–
1
of
1
results
1
Tocris Bioscience™ AC 264613

CAS: 1051487-82-1 Molecular Formula: C19H18BrN3O2 Molecular Weight (g/mol): 400.276