accessibility menu, dialog, popup

UserName

Uncover Lower Prices Daily with Price Drop

Sign In and Save

Viewing profile as user:
Fisher Scientific Logo
Don't have an account?  Create an Account
Main Menu
Admin
Main Menu
ULS Inventory Manager
Products

CAS RN 106819-03-8

CAS RN 106819-03-8

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.