CAS RN 108321-19-3
CAS RN 108321-19-3
IUPAC Name:
(2S)-N-(4-nitrophenyl)pyrrolidine-2-carboxamide; trifluoroacetic acid
Synonyms:
(2S)-N-(4-nitrophenyl)pyrrolidine-2-carboxamide; trifluoroacetic acid
Molecular Weight (g/mol):
349.27
Molecular Formula:
C13H14F3N3O5
InChi Key:
KYRVEVYREUUAKH-PPHPATTJSA-N
SMILES:
OC(=O)C(F)(F)F.[O-][N+](=O)C1=CC=C(NC(=O)[C@@H]2CCCN2)C=C1
1
–
1
of
1
results
1
Advanced Chem Tech H-Pro-pNA-TFA, Advanced ChemTech
CAS: 108321-19-3 Molecular Formula: C13H14F3N3O5 Molecular Weight (g/mol): 349.27 MDL Number: MFCD00069671 InChI Key: KYRVEVYREUUAKH-PPHPATTJSA-N IUPAC Name: (2S)-N-(4-nitrophenyl)pyrrolidine-2-carboxamide; trifluoroacetic acid SMILES: OC(=O)C(F)(F)F.[O-][N+](=O)C1=CC=C(NC(=O)[C@@H]2CCCN2)C=C1