accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108321-25-1

POHOOOHHOOH2N(R)

CAS RN 108321-25-1

IUPAC Name: (2R)-2-amino-3-[4-(phosphonooxy)phenyl]propanoic acid
Synonyms: O(4)-phospho-D-tyrosine
Molecular Weight (g/mol): 261.17
Molecular Formula: C9H12NO6P
InChi Key: DCWXELXMIBXGTH-MRVPVSSYSA-N
SMILES: N[C@H](CC1=CC=C(OP(O)(O)=O)C=C1)C(O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)