accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 108756-88-3

BHP

CAS RN 108756-88-3

IUPAC Name: dicyclohexylphosphane boron
Synonyms: dicyclohexylphosphane boron
Molecular Weight (g/mol): 209.1
Molecular Formula: C12H23BP
InChi Key: TVRLVVNHOYLIPR-UHFFFAOYSA-N
SMILES: [B].C1CCC(CC1)PC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results