CAS RN 109544-45-8
IUPAC Name:
2-(2-methoxy-2,3-dihydro-1,4-benzodioxin-2-yl)-4,5-dihydro-1H-imidazole hydrochloride
Synonyms:
2-(2-methoxy-3H-1,4-benzodioxin-2-yl)-4,5-dihydro-1H-imidazole hydrochloride
Molecular Weight (g/mol):
270.71
Molecular Formula:
C12H15ClN2O3
InChi Key:
IMPOOMVZVWKSAP-UHFFFAOYNA-N
SMILES:
Cl.COC1(COC2=CC=CC=C2O1)C1=NCCN1
1
–
1
of
1
results
1
R&D Systems™ RX 821002 hydrochloride
CAS: 109544-45-8 Molecular Formula: C12H15ClN2O3