accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 110637-40-6

OOHOOHNH2CHN

CAS RN 110637-40-6

IUPAC Name: 2-{[(prop-2-en-1-yloxy)carbonyl]amino}acetic acid; N-cyclohexylcyclohexanamine
Synonyms: dicha; {[(prop-2-en-1-yloxy)carbonyl]amino}acetic acid
Molecular Weight (g/mol): 340.46
Molecular Formula: C18H32N2O4
InChi Key: VVGNWCIOTXQIKJ-UHFFFAOYSA-N
SMILES: OC(=O)CNC(=O)OCC=C.C1CCC(CC1)NC1CCCCC1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)