CAS RN 111406-87-2
IUPAC Name:
1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea
Synonyms:
zileuton zyflo CR zyflo (TN) leutrol zyflo
Molecular Weight (g/mol):
236.29
Molecular Formula:
C11H12N2O2S
InChi Key:
MWLSOWXNZPKENC-UHFFFAOYNA-N
SMILES:
CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1
1
–
2
of
2
results
1
R&D Systems™ Zileuton
CAS: 111406-87-2 Molecular Formula: C11H12N2O2S
2
Thermo Scientific Chemicals Zileuton
CAS: 111406-87-2 Molecular Weight (g/mol): 236.29 MDL Number: MFCD00866097 InChI Key: MWLSOWXNZPKENC-UHFFFAOYNA-N IUPAC Name: 1-[1-(1-benzothiophen-2-yl)ethyl]-1-hydroxyurea SMILES: CC(N(O)C(N)=O)C1=CC2=CC=CC=C2S1