Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Zileuton
GREENER_CHOICESDP

Catalog No. 330850 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-085-0 50 mg
33-081-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 33-085-0 Supplier R&D Systems™ Supplier No. 3308/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. 5-LOX inhibitor; orally bioavailable

Orally active 5-lipoxygenase (5-LOX) inhibitor that inhibits LTB4 synthesis (IC50 values are 0.56, 2.3 and 2.6 μM in dog, rat and human blood respectively). Inhibits antigen-induced contraction of tracheal strips in vitro (IC50 = 6 μM) and exhibits antiasthmatic activity in vivo. Also weakly inhibits CYP1A2 (Ki = 66 - 98 μM).

Chemical Identifiers

CAS 111406-87-2
Molecular Formula C11H12N2O2S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 236.29
Purity >99%
Molecular Weight (g/mol) 236.29
Chemical Name or Material Zileuton
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.