CAS RN 112898-15-4
IUPAC Name:
({[({[5-(6-amino-9H-purin-9-yl)-3-(4-benzoylbenzoyloxy)-4-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid; triethylamine
Synonyms:
({[5-(6-aminopurin-9-yl)-3-(4-benzoylbenzoyloxy)-4-hydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxyphosphonic acid; triethylamine
Molecular Weight (g/mol):
816.59
Molecular Formula:
C30H39N6O15P3
InChi Key:
HVOVBTNCGADRTH-UHFFFAOYNA-N
SMILES:
CCN(CC)CC.NC1=C2N=CN(C3OC(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C(OC(=O)C4=CC=C(C=C4)C(=O)C4=CC=CC=C4)C3O)C2=NC=N1
1
–
1
of
1
results
1
R&D Systems™ BzATP triethylammonium salt
CAS: 112898-15-4 Molecular Formula: C30H39N6O15P3 Synonym: 2' 3'-o-4-benzoylbenzoyl adenosine 5'-triphosphate triethylammonium salt