CAS RN 112918-82-8
CAS RN 112918-82-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fmoc-Ida-OH, Advanced ChemTech
CAS: 112918-82-8 Molecular Formula: C19H17NO6 Molecular Weight (g/mol): 355.35 MDL Number: MFCD01321017 InChI Key: LNHILYINDTUUMZ-UHFFFAOYSA-N IUPAC Name: 2-[(carboxymethyl)({[(9H-fluoren-9-yl)methoxy]carbonyl})amino]acetic acid SMILES: OC(=O)CN(CC(O)=O)C(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2