CAS RN 1140525-25-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ SB 290157
A selective, high affinity, competitive antagonist of the anaphylatoxin C3a receptor. Does not antagonize C5aR or other chemotactic GPCRs.
2
Tocris Bioscience™ SB 290157 trifluoroacetate
Potent C3a receptor antagonist