accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON(R)NHSOOF

CAS RN 116649-85-5

IUPAC Name: 3-[(3R)-3-(4-fluorobenzenesulfonamido)-2,3,4,9-tetrahydro-1H-carbazol-9-yl]propanoic acid
Synonyms: ramatroban 雷马曲班 baynas
Molecular Weight (g/mol): 416.47
Molecular Formula: C21H21FN2O4S
InChi Key: LDXDSHIEDAPSSA-OAHLLOKOSA-N
SMILES: OC(=O)CCN1C2=C(C[C@@H](CC2)NS(=O)(=O)C2=CC=C(F)C=C2)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results