accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNONHNNSHCl

CAS RN 1186195-62-9

IUPAC Name: 4-(1-benzothiophen-2-yl)-N-{4-[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]phenyl}pyrimidin-2-amine hydrochloride
Synonyms: 4-(1-benzothiophen-2-yl)-N-{4-[4-(pyrrolidin-1-yl)piperidine-1-carbonyl]phenyl}pyrimidin-2-amine hydrochloride
Molecular Weight (g/mol): 520.09
Molecular Formula: C28H30ClN5OS
InChi Key: XFVDIVQCZWDBCW-UHFFFAOYSA-N
SMILES: Cl.O=C(N1CCC(CC1)N1CCCC1)C1=CC=C(NC2=NC=CC(=N2)C2=CC3=CC=CC=C3S2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)