accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNNHNH3COH3CONCH3CH3

CAS RN 1197196-48-7

IUPAC Name: 7-[3-(dimethylamino)propoxy]-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine
Synonyms: 7-[3-(dimethylamino)propoxy]-6-methoxy-2-(4-methyl-1,4-diazepan-1-yl)-N-(1-methylpiperidin-4-yl)quinazolin-4-amine
Molecular Weight (g/mol): 485.68
Molecular Formula: C26H43N7O2
InChi Key: XIVUGRBSBIXXJE-UHFFFAOYSA-N
SMILES: COC1=CC2=C(NC3CCN(C)CC3)N=C(N=C2C=C1OCCCN(C)C)N1CCCN(C)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)