CAS RN 121263-19-2
IUPAC Name:
1-[3,10-dihydroxy-12-(2-{[(4-hydroxyphenoxy)carbonyl]oxy}propyl)-2,6,7,11-tetramethoxy-4,9-dioxo-4,9-dihydroperylen-1-yl]propan-2-yl benzoate
Synonyms:
calphostin C
Molecular Weight (g/mol):
790.77
Molecular Formula:
C44H38O14
InChi Key:
LSUTUUOITDQYNO-UHFFFAOYNA-N
SMILES:
COC1=CC(=O)C2=C(O)C(OC)=C(CC(C)OC(=O)OC3=CC=C(O)C=C3)C3=C4C(CC(C)OC(=O)C5=CC=CC=C5)=C(OC)C(O)=C5C(=O)C=C(OC)C(C1=C23)=C45
1
–
2
of
2
results
1
R&D Systems™ Calphostin C
CAS: 121263-19-2 Molecular Formula: C44H38O14
2
MilliporeSigma™ Calbiochem™ Calphostin C, Cladosporium cladosporioides
A cell permeable, highly specific inhibitor of protein kinase C (IC50 = 50nM) that interacts with the protein's regulatory domain by competing at the binding site of diacylglycerol and phorbol esters.