Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Calphostin C
GREENER_CHOICESDP

Catalog No. 1626100U Shop All Novus Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-261-00U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 16-261-00U Supplier R&D Systems™ Supplier No. 1626/100U

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective and photo-dependent PKC inhibitor

Potent, selective and photo-dependent inhibitor of protein kinase C that targets the regulatory domain (IC50 = 50 nM). Displays > 1000-fold selectivity over other protein kinases such as cAMP-dependent protein kinase and tyrosine-specific protein kinase. Inhibits cell proliferation of malignant glioma cells in light-treated conditions in vitro (IC50 ∽ 40 - 60 nM). Also an antagonist of the Tcf/β-catenin complex.

Chemical Identifiers

CAS 121263-19-2
Molecular Formula C44H38O14

Specifications

Quantity 100 μg
Product Line Tocris
Formula Weight 790.76
Purity >98%
Molecular Weight (g/mol) 790.774
Chemical Name or Material Calphostin C
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.