accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1217457-86-7

IUPAC Name: methyl 2-({4-[(2-amino-3-sulfanylpropyl)amino]-2-(naphthalen-1-yl)phenyl}formamido)-4-methylpentanoate; trifluoroacetic acid
Synonyms: methyl 2-({4-[(2-amino-3-sulfanylpropyl)amino]-2-(naphthalen-1-yl)phenyl}formamido)-4-methylpentanoate; trifluoroacetic acid
Molecular Weight (g/mol): 593.66
Molecular Formula: C29H34F3N3O5S
InChi Key: WALKWJPZELDSKT-UHFFFAOYNA-N
SMILES: OC(=O)C(F)(F)F.COC(=O)C(CC(C)C)NC(=O)C1=C(C=C(NCC(N)CS)C=C1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results