CAS RN 1217466-21-1
IUPAC Name:
2-[(2-benzoylphenyl)amino]-3-(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)propanoic acid hydrochloride
Synonyms:
2-[(2-benzoylphenyl)amino]-3-(4-{2-[methyl(pyridin-2-yl)amino]ethoxy}phenyl)propanoic acid hydrochloride
Molecular Weight (g/mol):
532.04
Molecular Formula:
C30H30ClN3O4
InChi Key:
KXNKIKXTGRMLEY-UHFFFAOYNA-N
SMILES:
Cl.CN(CCOC1=CC=C(CC(NC2=CC=CC=C2C(=O)C2=CC=CC=C2)C(O)=O)C=C1)C1=CC=CC=N1
1
–
1
of
1
results
1
R&D Systems™ GW 1929 hydrochloride
CAS: 1217466-21-1 Molecular Formula: C30H30ClN3O4