CAS RN 1234480-84-2
IUPAC Name:
5-({2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Synonyms:
5-({2-methoxy-4-[4-(4-methylpiperazin-1-yl)piperidine-1-carbonyl]phenyl}amino)-2,9-dimethyl-2,4,6,9-tetraazatricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,11,13-hexaen-10-one
Molecular Weight (g/mol):
570.7
Molecular Formula:
C31H38N8O3
InChi Key:
IWMCPJZTADUIFX-UHFFFAOYSA-N
SMILES:
COC1=C(NC2=NC=C3N(C)C(=O)C4=CC=CC=C4N(C)C3=N2)C=CC(=C1)C(=O)N1CCC(CC1)N1CCN(C)CC1