accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOFFFHOCH3H3CCH3

CAS RN 123557-91-5

IUPAC Name: 5,7-di-tert-butyl-3-hydroxy-3-(trifluoromethyl)-2,3-dihydro-1-benzofuran-2-one
Synonyms: BHFF
Molecular Weight (g/mol): 330.35
Molecular Formula: C17H21F3O3
InChi Key: RVNOANDLZIIFHB-UHFFFAOYNA-N
SMILES: CC(C)(C)C1=CC(=C2OC(=O)C(O)(C2=C1)C(F)(F)F)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)