accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNOOOHNOO(E/Z)

CAS RN 1237744-13-6

IUPAC Name: 4-{6-methoxy-2-[2-(3-nitrophenyl)ethenyl]-4-oxo-3,4-dihydroquinazolin-3-yl}benzoic acid
Synonyms: 4-{6-methoxy-2-[2-(3-nitrophenyl)ethenyl]-4-oxoquinazolin-3-yl}benzoic acid
Molecular Weight (g/mol): 443.42
Molecular Formula: C24H17N3O6
InChi Key: GNLVJIICVWDSNI-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(C=CC3=CC(=CC=C3)[N+]([O-])=O)N(C3=CC=C(C=C3)C(O)=O)C(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)