CAS RN 124431-80-7
IUPAC Name:
3-{4-[2-({6-amino-9-[5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]-9H-purin-2-yl}amino)ethyl]phenyl}propanoic acid hydrochloride
Synonyms:
3-{4-[2-({6-amino-9-[5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl}amino)ethyl]phenyl}propanoic acid hydrochloride
Molecular Weight (g/mol):
535.99
Molecular Formula:
C23H30ClN7O6
InChi Key:
QPHVMNOEKKJYJO-UHFFFAOYNA-N
SMILES:
Cl.CCNC(=O)C1OC(C(O)C1O)N1C=NC2=C(N)N=C(NCCC3=CC=C(CCC(O)=O)C=C3)N=C12
1
–
1
of
1
results
1
R&D Systems™ CGS 21680 hydrochloride
CAS: 124431-80-7 Molecular Formula: C23H30ClN7O6